siRNA Details
| Gene Symbol: Fgf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008006.2 | NP_032032.1 | 3 | 549 | ||
| GenBank mRNA | AF065903.1 | - | 3 | 352 | ||
| AF065904.1 | - | 3 | 352 | |||
| AF065905.1 | - | 3 | 352 | |||
| AY027548.1 | - | 3 | 267 | |||
| AY027549.1 | - | 3 | 372 | |||
| AY027550.1 | - | 1 | 144 | |||
| AY027551.1 | - | 3 | 349 | |||
| AY027557.1 | - | 3 | 422 | |||
| AY027558.1 | - | 3 | 413 | |||
| AY027559.1 | - | 3 | 276 | |||
| M30644.1 | - | 3 | 352 | |||

Fgf2
Genomic Map