siRNA Details
| Gene Symbol: Fgf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010202.5 | NP_034332.2 | 3 | 691 | ||
| GenBank mRNA | AK082814.1 | - | 3 | 692 | ||
| BC104312.1 | - | 3 | 572 | 40044462 | ||
| BC104313.1 | - | 3 | 572 | 40044466 | ||
| M30642.1 | - | 3 | 572 | |||

Fgf4
Genomic Map