siRNA Details
| Gene Symbol: Fgr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010208.4 | NP_034338.3 | 3 | 401 | ||
| GenBank mRNA | AK036438.1 | - | 3 | 323 | ||
| AK036476.1 | - | 3 | 573 | |||
| AK038141.1 | - | 3 | 307 | |||
| AK089812.1 | - | 3 | 371 | |||
| AK155902.1 | - | 3 | 377 | |||
| AK157640.1 | - | 355 | ||||
| AK171824.1 | - | 3 | 355 | |||
| BC057863.1 | - | 3 | 270 | 5317449 | ||
| X16440.1 | - | 3 | 336 | |||
| X52191.1 | - | 3 | 308 | |||

Fgr
Genomic Map