siRNA Details
Gene Symbol: Fhl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010213.3 | NP_034343.2 | 5 | 824 | ||
GenBank mRNA | AF114382.1 | - | 5 | 795 | ||
AF149826.1 | - | 4 | 622 | |||
BC144903.1 | - | 5 | 788 | 9053451 | ||
BC145939.1 | - | 5 | 788 | 40131242 | ||
CB205184.1 | - | 3 | 481 |
