siRNA Details
| Gene Symbol: Fkbp10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163481.1 | NP_001156953.1 | 10 | 1672 | ||
| NM_010221.2 | NP_034351.2 | 10 | 1675 | |||
| GenBank mRNA | AK019446.2 | - | 10 | 1614 | ||
| AK144185.1 | - | 10 | 1655 | |||
| BC029546.1 | - | 10 | 1622 | 5059670 | ||
| L07063.1 | - | 10 | 1678 | |||

Fkbp10
Genomic Map