siRNA Details
| Gene Symbol: Flt3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013520.3 | NP_038548.3 | 3, 4 | 238 | ||
| GenBank mRNA | AK020105.1 | - | 3, 4 | 238 | ||
| BC019801.1 | - | 3, 4 | 223 | 5132319 | ||
| CK622267.1 | - | 3, 4 | 208 | |||
| EU274583.1 | - | 2, 3 | 150 | |||
| L23636.1 | - | 2, 3 | 181 | |||
| S76459.1 | - | 2, 3 | 156 | |||
| U04807.1 | - | 2, 3 | 405 | |||

Flt3l
Genomic Map