siRNA Details
| Gene Symbol: Asb13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001267724.1 | NP_001254653.1 | 4 | 408 | ||
| NM_178283.4 | NP_840068.1 | 6 | 883 | |||
| GenBank mRNA | AF403041.1 | - | 6 | 883 | ||
| AK078289.1 | - | 6 | 839 | |||
| AK133892.1 | - | 6 | 807 | |||
| BC018240.1 | - | 6 | 837 | 4239996 | ||
| BG080214.2 | - | 4 | 342 | |||

Asb13
Genomic Map