siRNA Details
| Gene Symbol: Fnbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001038700.2 | NP_001033789.1 | 2, 3 | 344 | ||
| NM_001177648.1 | NP_001171119.1 | 2, 3 | 344 | |||
| NM_001177649.1 | NP_001171120.1 | 2, 3 | 344 | |||
| NM_019406.3 | NP_062279.1 | 2, 3 | 344 | |||
| GenBank mRNA | AK122308.1 | - | 2, 3 | 344 | ||
| AK157135.1 | - | 2, 3 | 335 | |||
| AK170497.1 | - | 2, 3 | 294 | |||
| AK172100.1 | - | 2, 3 | 288 | |||
| BC003867.1 | - | 2, 3 | 275 | 3490113 | ||

Fnbp1
Genomic Map