siRNA Details
| Gene Symbol: Fnbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001038700.2 | NP_001033789.1 | 8 | 953 | ||
| NM_001177648.1 | NP_001171119.1 | 8 | 953 | |||
| NM_001177649.1 | NP_001171120.1 | 8 | 953 | |||
| NM_001177650.1 | NP_001171121.1 | 6 | 650 | |||
| NM_019406.3 | NP_062279.1 | 8 | 953 | |||
| GenBank mRNA | AK052652.1 | - | 2 | 245 | ||
| AK122308.1 | - | 8 | 953 | |||
| AK143512.1 | - | 6 | 651 | |||
| AK157135.1 | - | 8 | 944 | |||
| AK170497.1 | - | 8 | 903 | |||
| AK172100.1 | - | 8 | 897 | |||
| BC003867.1 | - | 8 | 884 | 3490113 | ||

Fnbp1
Genomic Map