siRNA Details
| Gene Symbol: Fzd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021457.3 | NP_067432.2 | 1 | 2336 | ||
| GenBank mRNA | AF054623.2 | - | 1 | 1700 | ||
| AK143101.1 | - | 1 | 2156 | |||
| BC053010.1 | - | 1 | 2091 | 5697795 | ||
More Information
Other Species Targeted
Set Membership: |
|

Fzd1
Genomic Map