siRNA Details
| Gene Symbol: G0s2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008059.3 | NP_032085.1 | 2 | 345 | ||
| GenBank mRNA | AK003165.1 | - | 2 | 267 | ||
| BC117038.1 | - | 2 | 173 | 40126357 | ||
| BC117040.1 | - | 2 | 173 | 40126359 | ||
| X95280.1 | - | 2 | 249 | |||

G0s2
Genomic Map