siRNA Details
| Gene Symbol: Gaa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159324.1 | NP_001152796.1 | 13 | 1960 | ||
| NM_008064.3 | NP_032090.3 | 13 | 2050 | |||
| GenBank mRNA | AK052211.1 | - | 13 | 2031 | ||
| AK088481.1 | - | 13 | 2026 | |||
| AK139333.1 | - | 13 | 1998 | |||
| AK140042.1 | - | 13 | 2018 | |||
| AK141546.1 | - | 13 | 2033 | |||
| AK145631.1 | - | 12 | 1466 | |||
| AK146538.1 | - | 13 | 1954 | |||
| AK150970.1 | - | 13 | 2008 | |||
| BC010210.1 | - | 13 | 2014 | 3969127 | ||
| U49351.1 | - | 13 | 1902 | |||

Gaa
Genomic Map