siRNA Details
| Gene Symbol: Gaa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159324.1 | NP_001152796.1 | 2, 3 | 723 | ||
| NM_008064.3 | NP_032090.3 | 2, 3 | 813 | |||
| GenBank mRNA | AK052211.1 | - | 2, 3 | 794 | ||
| AK054057.1 | - | 2, 3 | 794 | |||
| AK088481.1 | - | 2, 3 | 789 | |||
| AK139333.1 | - | 2, 3 | 761 | |||
| AK140042.1 | - | 2, 3 | 779 | |||
| AK141546.1 | - | 2, 3 | 796 | |||
| AK145631.1 | - | 1, 2 | 228 | |||
| AK146538.1 | - | 2, 3 | 717 | |||
| AK150970.1 | - | 2, 3 | 771 | |||
| BC010210.1 | - | 2, 3 | 777 | 3969127 | ||

Gaa
Genomic Map