siRNA Details
| Gene Symbol: Slc6a12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133661.3 | NP_598422.2 | 12 | 1691 | ||
| GenBank mRNA | BC019211.1 | - | 12 | 1695 | 5053116 | |
| M97632.1 | - | 12 | 1431 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc6a12| Gene Symbol: Slc6a12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133661.3 | NP_598422.2 | 12 | 1691 | ||
| GenBank mRNA | BC019211.1 | - | 12 | 1695 | 5053116 | |
| M97632.1 | - | 12 | 1431 | |||