siRNA Details
| Gene Symbol: Galnt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015736.2 | NP_056551.2 | 2 | 666 | ||
| GenBank mRNA | AK135489.1 | - | 2 | 627 | ||
| BC043331.1 | - | 2 | 522 | 5342768 | ||
| U70538.1 | - | 2 | 510 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Galnt3| Gene Symbol: Galnt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015736.2 | NP_056551.2 | 2 | 666 | ||
| GenBank mRNA | AK135489.1 | - | 2 | 627 | ||
| BC043331.1 | - | 2 | 522 | 5342768 | ||
| U70538.1 | - | 2 | 510 | |||