siRNA Details
| Gene Symbol: Gbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010260.1 | NP_034390.1 | 8 | 1348 | ||
| GenBank mRNA | AF077007.1 | - | 8 | 1348 | ||
| AF109168.1 | - | 8 | 1301 | |||
| AJ007970.1 | - | 8 | 1341 | |||
| AK150138.1 | - | 8 | 1284 | |||
| AK150146.1 | - | 4 | 650 | |||
| BC011336.1 | - | 8 | 1337 | 3494311 | ||
| BC032882.1 | - | 8 | 1363 | 1230883 | ||
| BC049141.1 | - | 8 | 1325 | |||
| CT010249.1 | - | 7 | 1178 | |||

Gbp2
Genomic Map