siRNA Details
| Gene Symbol: Gbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010260.1 | NP_034390.1 | 9 | 1580 | ||
| GenBank mRNA | AF077007.1 | - | 9 | 1580 | ||
| AF109168.1 | - | 9 | 1533 | |||
| AJ007970.1 | - | 9 | 1573 | |||
| AK150138.1 | - | 9 | 1516 | |||
| AK150146.1 | - | 5 | 882 | |||
| BC011336.1 | - | 9 | 1569 | 3494311 | ||
| BC032882.1 | - | 9 | 1595 | 1230883 | ||
| BC049141.1 | - | 9 | 1557 | |||
| CT010249.1 | - | 8 | 1410 | |||

Gbp2
Genomic Map