siRNA Details
| Gene Symbol: Gdf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008108.4 | NP_032134.2 | 2 | 426 | ||
| GenBank mRNA | AK082877.1 | - | 2 | 347 | ||
| AK146174.1 | - | 2 | 352 | |||
| AK152457.1 | - | 2 | 427 | |||
| AK160533.1 | - | 2 | 384 | |||
| AK167962.1 | - | 2 | 352 | |||
| BC101963.1 | - | 2 | 341 | 40045099 | ||
| BC101964.1 | - | 2 | 341 | 40045100 | ||
| BC101965.1 | - | 2 | 342 | |||
| BC103565.1 | - | 2 | 340 | 40045101 | ||
| L06443.1 | - | 2 | 442 | |||
| S52658.1 | - | 2 | 382 | |||

Gdf3
Genomic Map