siRNA Details
Gene Symbol: Gfap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001131020.1 | NP_001124492.1 | 1 | 316 | ||
NM_010277.3 | NP_034407.2 | 1 | 316 | |||
GenBank mRNA | AF332061.1 | - | 1 | 143 | ||
AF332062.1 | - | 1 | 143 | |||
AK079577.1 | - | 1 | 250 | |||
AK140151.1 | - | 1 | 248 | |||
BC100728.1 | - | 1 | 195 | |||
BC100737.1 | - | 1 | 188 | |||
BC101968.1 | - | 1 | 195 | |||
BC103571.1 | - | 1 | 189 | |||
BC139357.1 | - | 1 | 316 | 8862379 | ||
BC139358.1 | - | 1 | 316 | 8862380 | ||
K01347.1 | - | 1 | 152 | |||
M25937.1 | - | 1 | 152 |
