siRNA Details
| Gene Symbol: Gfra1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001285457.1 | NP_001272386.1 | 8 | 1450 | ||
| NM_010279.3 | NP_034409.1 | 10 | 1666 | |||
| GenBank mRNA | AB000800.1 | - | 9 | 1220 | ||
| AF012811.1 | - | 5 | 492 | |||
| AF015172.1 | - | 8 | 1433 | |||
| AK076171.1 | - | 10 | 1587 | |||
| AK139987.1 | - | 5 | 507 | |||
| BC011532.1 | - | 8 | 1397 | |||
| BC040251.1 | - | 8 | 1234 | |||
| BC044783.2 | - | 11 | 1632 | |||
| BC054378.1 | - | 9 | 1337 | 6390018 | ||

Gfra1
Genomic Map