siRNA Details
| Gene Symbol: Ghr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001048178.2 | NP_001041643.1 | 5, 6 | 642 | ||
| NM_001286370.1 | NP_001273299.1 | 5, 6 | 343 | |||
| NM_010284.3 | NP_034414.2 | 5, 6 | 642 | |||
| GenBank mRNA | AK143886.1 | - | 5, 6 | 344 | ||
| AK143925.1 | - | 5, 6 | 440 | |||
| BC024375.1 | - | 5, 6 | 477 | |||
| BC056645.1 | - | 5, 6 | 458 | |||
| BC075720.1 | - | 5, 6 | 490 | 5044060 | ||
| M31680.1 | - | 5, 6 | 378 | |||
| M33324.1 | - | 5, 6 | 378 | |||

Ghr
Genomic Map