siRNA Details
| Gene Symbol: Gjb6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010937.2 | NP_001010937.1 | 3 | 1322 | ||
| NM_001271663.1 | NP_001258592.1 | 3 | 1413 | |||
| GenBank mRNA | AK134617.1 | - | 3 | 1321 | ||
| AK141610.1 | - | 3 | 1322 | |||
| BC013811.1 | - | 3 | 989 | 3483477 | ||
| BC016507.1 | - | 3 | 997 | 3481872 | ||
| BC063762.1 | - | 1 | 1136 | 30245263 | ||

Gjb6
Genomic Map