siRNA Details
| Gene Symbol: Glul | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008131.4 | NP_032157.2 | 6 | 893 | ||
| GenBank mRNA | AK137969.1 | - | 6 | 880 | ||
| AK159106.1 | - | 6 | 879 | |||
| AK160670.1 | - | 6 | 880 | |||
| AK162685.1 | - | 6 | 878 | |||
| AK168493.1 | - | 6 | 877 | |||
| AY044241.1 | - | 6 | 877 | |||
| BC015086.1 | - | 6 | 877 | 4224865 | ||

Glul
Genomic Map