siRNA Details
| Gene Symbol: Gnai2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008138.4 | NP_032164.2 | 4 | 466 | ||
| GenBank mRNA | AK157998.1 | - | 4 | 470 | ||
| AK159222.1 | - | 4 | 470 | |||
| AK160186.1 | - | 4 | 462 | |||
| AK167388.1 | - | 4 | 468 | |||
| AK185434.1 | - | 1 | 40 | |||
| AK185480.1 | - | 365 | ||||
| AK202003.1 | - | 4 | 346 | |||
| BC065159.1 | - | 4 | 453 | 5717305 | ||
| M13963.1 | - | 4 | 438 | |||

Gnai2
Genomic Map