siRNA Details
| Gene Symbol: Gnal | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010307.3 | NP_034437.1 | 10 | 1084 | ||
| NM_177137.5 | NP_796111.2 | 10 | 1249 | |||
| GenBank mRNA | AK158831.1 | - | 10 | 1085 | ||
| AY179169.1 | - | 10 | 818 | |||
| BC078439.1 | - | 10 | 1211 | 5685967 | ||
| U38503.1 | - | 3 | 194 | |||

Gnal
Genomic Map