siRNA Details
| Gene Symbol: Gnat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008141.3 | NP_032167.1 | 9 | 972 | ||
| GenBank mRNA | AK009388.1 | - | 8 | 856 | ||
| AK028395.1 | - | 5 | 489 | |||
| AY974584.1 | - | 2 | 125 | |||
| BC016272.1 | - | 9 | 970 | 4506728 | ||
| BC085102.1 | - | 7 | 1719 | |||
| BC131941.1 | - | 9 | 967 | 40130218 | ||
| BC131943.1 | - | 9 | 967 | 40130220 | ||
| L10666.1 | - | 8 | 894 | |||

Gnat2
Genomic Map