siRNA Details
| Gene Symbol: Gnaz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010311.3 | NP_034441.1 | 4 | 1539 | ||
| GenBank mRNA | AF056972.1 | - | 2 | 474 | ||
| AK080716.1 | - | 4 | 1539 | |||
| AK133483.1 | - | 2 | 1415 | |||
| AK134710.1 | - | 2 | 1243 | |||
| BC014702.1 | - | 2 | 1240 | 4501587 | ||

Gnaz
Genomic Map