siRNA Details
| Gene Symbol: Lancl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190984.1 | NP_001177913.1 | 10 | 1259 | ||
| NM_001190985.1 | NP_001177914.1 | 9 | 1477 | |||
| NM_021295.3 | NP_067270.1 | 10 | 1293 | |||
| GenBank mRNA | AJ294535.1 | - | 10 | 1286 | ||
| AK146120.1 | - | 9 | 1468 | |||
| BC038649.1 | - | 7 | 709 | |||
| BC058560.1 | - | 10 | 1259 | 6315377 | ||
| Y11550.1 | - | 10 | 1258 | |||
| Y16518.1 | - | 9 | 1437 | |||

Lancl1
Genomic Map