siRNA Details
| Gene Symbol: Lancl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190984.1 | NP_001177913.1 | 4, 5 | 490 | ||
| NM_001190985.1 | NP_001177914.1 | 3, 4 | 708 | |||
| NM_021295.3 | NP_067270.1 | 4, 5 | 524 | |||
| GenBank mRNA | AJ294535.1 | - | 4, 5 | 517 | ||
| AK208199.1 | - | 1, 2 | 80 | |||
| BC038649.1 | - | 1, 2 | 29 | |||
| BC058560.1 | - | 4, 5 | 490 | 6315377 | ||
| Y11550.1 | - | 4, 5 | 489 | |||
| Y16518.1 | - | 3, 4 | 668 | |||

Lancl1
Genomic Map