siRNA Details
| Gene Symbol: Grk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080743.1 | NP_001074212.1 | 5 | 958 | ||
| NM_019497.2 | NP_062370.2 | 5 | 958 | |||
| GenBank mRNA | AF040745.1 | - | 5 | 384 | ||
| AK020792.1 | - | 5 | 958 | |||
| AK078475.1 | - | 6 | 830 | |||
| AK132685.1 | - | 5 | 959 | |||
| BC150691.1 | - | 5 | 482 | 9087602 | ||

Grk4
Genomic Map