siRNA Details
| Gene Symbol: Gria1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113325.2 | NP_001106796.1 | 1 | 464 | ||
| NM_008165.4 | NP_032191.2 | 1 | 464 | |||
| GenBank mRNA | AB102773.1 | - | 1 | 48 | ||
| AB102774.1 | - | 1 | 48 | |||
| AK049958.1 | - | 1 | 274 | |||
| BC056397.1 | - | 1 | 126 | 6842391 | ||
| BC060702.1 | - | 1 | 47 | |||
| CJ155313.1 | - | 1 | 392 | |||
| JN180918.1 | - | 115 | ||||
| X57497.1 | - | 1 | 311 | |||

Gria1
Genomic Map