siRNA Details
Gene Symbol: Gria1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113325.2 | NP_001106796.1 | 2 | 512 | ||
NM_001252403.1 | NP_001239332.1 | 2 | 139 | |||
NM_008165.4 | NP_032191.2 | 2 | 512 | |||
GenBank mRNA | AB102773.1 | - | 2 | 96 | ||
AB102774.1 | - | 2 | 96 | |||
AK049958.1 | - | 2 | 322 | |||
AK135107.1 | - | 2 | 140 | |||
BC056397.1 | - | 2 | 174 | 6842391 | ||
BC060702.1 | - | 2 | 95 | |||
CJ155313.1 | - | 2 | 440 | |||
JN180918.1 | - | 163 | ||||
JN180919.1 | - | 2 | 94 | |||
X57497.1 | - | 2 | 359 |
