siRNA Details
| Gene Symbol: Grik2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111268.1 | NP_001104738.1 | 8 | 1438 | ||
| NM_010349.2 | NP_034479.2 | 8 | 1438 | |||
| GenBank mRNA | BC131639.1 | - | 7 | 1119 | ||
| BC131640.1 | - | 7 | 1119 | |||
| BC148311.2 | - | 7 | 1119 | |||
| D10054.1 | - | 8 | 1351 | |||
| X66117.1 | - | 7 | 1175 | |||

Grik2
Genomic Map