siRNA Details
| Gene Symbol: Grik2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111268.1 | NP_001104738.1 | 5 | 998 | ||
| NM_010349.2 | NP_034479.2 | 5 | 998 | |||
| GenBank mRNA | AK141539.1 | - | 5 | 999 | ||
| BC060154.1 | - | 4 | 793 | |||
| BC131639.1 | - | 4 | 679 | |||
| BC131640.1 | - | 4 | 679 | |||
| BC148311.2 | - | 4 | 679 | |||
| D10054.1 | - | 5 | 911 | |||
| X66117.1 | - | 4 | 735 | |||

Grik2
Genomic Map