siRNA Details
| Gene Symbol: Gsg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010353.2 | NP_034483.1 | 1 | 1153 | ||
| GenBank mRNA | BC113148.1 | - | 1 | 1147 | 40054435 | |
| D87326.1 | - | 1 | 1153 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gsg2| Gene Symbol: Gsg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010353.2 | NP_034483.1 | 1 | 1153 | ||
| GenBank mRNA | BC113148.1 | - | 1 | 1147 | 40054435 | |
| D87326.1 | - | 1 | 1153 | |||