siRNA Details
| Gene Symbol: Gsx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008178.2 | NP_032204.1 | 2 | 872 | ||
| GenBank mRNA | AB221618.1 | - | 2 | 682 | ||
| BC137769.1 | - | 2 | 841 | 8860788 | ||
| U21224.1 | - | 2 | 872 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gsx1| Gene Symbol: Gsx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008178.2 | NP_032204.1 | 2 | 872 | ||
| GenBank mRNA | AB221618.1 | - | 2 | 682 | ||
| BC137769.1 | - | 2 | 841 | 8860788 | ||
| U21224.1 | - | 2 | 872 | |||