siRNA Details
| Gene Symbol: Gstm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010358.5 | NP_034488.1 | 8 | 811 | ||
| GenBank mRNA | BC003822.1 | - | 8 | 634 | 3600444 | |
| BC046758.1 | - | 8 | 664 | 5693552 | ||
| BC091763.1 | - | 8 | 644 | 5102093 | ||
| J03952.1 | - | 8 | 625 | |||
| J04632.1 | - | 8 | 812 | |||

Gstm1
Genomic Map