siRNA Details
| Gene Symbol: Cfap20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008187.2 | NP_032213.2 | 1 | 322 | ||
| GenBank mRNA | AK088713.1 | - | 1 | 226 | ||
| AK172603.1 | - | 1 | 298 | |||
| BC092397.1 | - | 1 | 210 | |||
| BC119023.1 | - | 1 | 218 | 8733772 | ||
| BC119025.1 | - | 1 | 218 | 8733774 | ||
| BC144770.1 | - | 1 | 218 | |||
| BM934726.1 | - | 1 | 322 | |||
| Z54179.1 | - | 1 | 245 | |||

Cfap20
Genomic Map