siRNA Details
| Gene Symbol: Trip12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133975.4 | NP_598736.4 | 11, 12 | 1834 | ||
| GenBank mRNA | AK134397.1 | - | 11, 12 | 1793 | ||
| AK158807.1 | - | 11, 12 | 1834 | |||
| BC044869.1 | - | 8, 9 | 792 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trip12| Gene Symbol: Trip12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133975.4 | NP_598736.4 | 11, 12 | 1834 | ||
| GenBank mRNA | AK134397.1 | - | 11, 12 | 1793 | ||
| AK158807.1 | - | 11, 12 | 1834 | |||
| BC044869.1 | - | 8, 9 | 792 | |||