siRNA Details
| Gene Symbol: Nkx6-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183248.3 | NP_899071.2 | 1 | 524 | ||
| GenBank mRNA | AK008173.1 | - | 1 | 524 | ||
| L08074.1 | - | 1 | 462 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nkx6-2| Gene Symbol: Nkx6-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183248.3 | NP_899071.2 | 1 | 524 | ||
| GenBank mRNA | AK008173.1 | - | 1 | 524 | ||
| L08074.1 | - | 1 | 462 | |||