siRNA Details
| Gene Symbol: Gzmg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010375.2 | NP_034505.1 | 5 | 701 | ||
| GenBank mRNA | BC099473.1 | - | 5 | 701 | 30715974 | |
| J02872.1 | - | 5 | 687 | |||
| M36900.1 | - | 4 | 496 | |||
| X14092.1 | - | 4 | 496 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gzmg| Gene Symbol: Gzmg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010375.2 | NP_034505.1 | 5 | 701 | ||
| GenBank mRNA | BC099473.1 | - | 5 | 701 | 30715974 | |
| J02872.1 | - | 5 | 687 | |||
| M36900.1 | - | 4 | 496 | |||
| X14092.1 | - | 4 | 496 | |||