siRNA Details
| Gene Symbol: H1f0 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008197.3 | NP_032223.2 | 1 | 892 | ||
| GenBank mRNA | AK051213.1 | - | 1 | 655 | ||
| AK077473.1 | - | 1 | 569 | |||
| AK090029.1 | - | 1 | 657 | |||
| AK145675.1 | - | 1 | 544 | |||
| AK146077.1 | - | 1 | 655 | |||
| AK153990.1 | - | 1 | 656 | |||
| AK155312.1 | - | 1 | 587 | |||
| AK177351.1 | - | 1 | 150 | |||
| AK186840.1 | - | 1 | 150 | |||
| BC003830.1 | - | 1 | 641 | 3600538 | ||
| BC011493.1 | - | 1 | 639 | 4166167 | ||
| BC110361.1 | - | 1 | 644 | 5343233 | ||
| X13171.1 | - | 1 | 622 | |||

H1f0
Genomic Map