siRNA Details
| Gene Symbol: Ifi44l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031367.1 | NP_112735.1 | 1366 | |||
| GenBank mRNA | AF336221.1 | - | 1366 | |||
| BC024930.1 | - | 1237 | 4020690 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ifi44l| Gene Symbol: Ifi44l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031367.1 | NP_112735.1 | 1366 | |||
| GenBank mRNA | AF336221.1 | - | 1366 | |||
| BC024930.1 | - | 1237 | 4020690 | |||