siRNA Details
| Gene Symbol: Hap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010404.3 | NP_034534.1 | 11 | 1844 | ||
| GenBank mRNA | AJ002271.1 | - | 11 | 1790 | ||
| AK138436.1 | - | 1794 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hap1| Gene Symbol: Hap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010404.3 | NP_034534.1 | 11 | 1844 | ||
| GenBank mRNA | AJ002271.1 | - | 11 | 1790 | ||
| AK138436.1 | - | 1794 | ||||