siRNA Details
| Gene Symbol: Hba-x | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010405.4 | NP_034535.1 | 3 | 392 | ||
| GenBank mRNA | AK077590.1 | - | 3 | 391 | ||
| BC051988.1 | - | 3 | 362 | 5711898 | ||
| M13125.1 | - | 2 | 200 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hba-x| Gene Symbol: Hba-x | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010405.4 | NP_034535.1 | 3 | 392 | ||
| GenBank mRNA | AK077590.1 | - | 3 | 391 | ||
| BC051988.1 | - | 3 | 362 | 5711898 | ||
| M13125.1 | - | 2 | 200 | |||