siRNA Details
| Gene Symbol: Foxf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010426.2 | NP_034556.2 | 1, 2 | 988 | ||
| GenBank mRNA | BC138805.1 | - | 1, 2 | 988 | 8861827 | |
| BC138806.1 | - | 1, 2 | 988 | 8861828 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Foxf1| Gene Symbol: Foxf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010426.2 | NP_034556.2 | 1, 2 | 988 | ||
| GenBank mRNA | BC138805.1 | - | 1, 2 | 988 | 8861827 | |
| BC138806.1 | - | 1, 2 | 988 | 8861828 | ||