siRNA Details
| Gene Symbol: Tfb2m | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008249.4 | NP_032275.2 | 4, 5 | 842 | ||
| GenBank mRNA | AK090106.1 | - | 4, 5 | 813 | ||
| AK146563.1 | - | 4, 5 | 843 | |||
| AK166717.1 | - | 4, 5 | 829 | |||
| AK167930.1 | - | 4, 5 | 794 | |||
| AK168970.1 | - | 4, 5 | 821 | |||
| AK169138.1 | - | 4, 5 | 821 | |||
| BC138836.1 | - | 4, 5 | 787 | 8861858 | ||
| BC138837.1 | - | 4, 5 | 787 | 8861859 | ||
| M74555.1 | - | 4, 5 | 793 | |||

Tfb2m
Genomic Map