siRNA Details
| Gene Symbol: Hmox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136066.2 | NP_001129538.1 | 4 | 362 | ||
| NM_010443.2 | NP_034573.2 | 4 | 378 | |||
| GenBank mRNA | AF029874.1 | - | 4 | 375 | ||
| AF054669.1 | - | 4 | 379 | |||
| AF054670.1 | - | 4 | 313 | |||
| AK032175.1 | - | 4 | 320 | |||
| AK134536.1 | - | 4 | 337 | |||
| AK141426.1 | - | 4 | 356 | |||
| AK152936.1 | - | 4 | 301 | |||
| BC002011.1 | - | 4 | 334 | 3484259 | ||
| BY071995.1 | - | 2 | 125 | |||
| CA465845.1 | - | 4 | 393 | |||
| Gene Symbol: Cdip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC029975.1 | - | 35 | |||

Hmox2
Genomic Map