siRNA Details
Gene Symbol: Hmx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145998.3 | NP_666110.1 | 2 | 1049 | ||
GenBank mRNA | AK053014.1 | - | 2 | 819 | ||
AK138228.1 | - | 2 | 1050 | |||
BC023402.1 | - | 3 | 919 | 5043712 | ||
S80989.1 | - | 2 | 843 |
