siRNA Details
| Gene Symbol: Hmx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008257.3 | NP_032283.3 | 2 | 1008 | ||
| GenBank mRNA | AI326757.1 | - | 1 | 361 | ||
| BC139482.1 | - | 2 | 1104 | 8862504 | ||
| BC139483.1 | - | 2 | 1104 | 8862505 | ||
| BE649795.1 | - | 1 | 275 | |||

Hmx3
Genomic Map